Chisels, Aesculap®
Chisel, Partsch, 170 mm, 7 mm
Katalog # AESCFL187R
Leverandør: Aesculap
Gå til:
Product Details & Documents
These chisels are used for dental surgery.
Specifikationer
- Beskrivelse:Chisel
- Længde:170 mm
- Seq:13
- Model:Partsch
- Bredde:7 mm