Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Chisels, Aesculap®

Chisel, Partsch, 170 mm, 3 mm
Katalog # AESCFL183R
Leverandør:   Aesculap
Chisels, Aesculap®
Katalog # AESCFL183R
Leverandør:  Aesculap
Chisels, Aesculap®

Chisel, Partsch, 170 mm, 3 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These chisels are used for dental surgery.

Specifikationer
  • Beskrivelse:
    Chisel
  • Længde:
    170 mm
  • Seq:
    09
  • Model:
    Partsch
  • Bredde:
    3 mm