Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Chisels, Aesculap®

Chisels, Aesculap®

Chisel, Partsch, 140 mm, 6 mm
Katalog # AESCFL176R
Leverandør:   Aesculap
Chisels, Aesculap®
Katalog # AESCFL176R
Leverandør:  Aesculap
Chisels, Aesculap®

Chisel, Partsch, 140 mm, 6 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These chisels are used for dental surgery.

Specifikationer
  • Beskrivelse:
    Chisel
  • Længde:
    140 mm
  • Seq:
    08
  • Model:
    Partsch
  • Bredde:
    6 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.