Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Chisels, Aesculap®

Chisel, Partsch, 140 mm, 5 mm
  AESCFL175R
 :   Aesculap
Chisels, Aesculap®
  AESCFL175R
 :  Aesculap
Chisels, Aesculap®

Chisel, Partsch, 140 mm, 5 mm

 
 
   
1 * 1 Items
 

These chisels are used for dental surgery.

 
  • Beskrivelse:
    Chisel
  • Længde:
    140 mm
  • Seq:
    07
  • Model:
    Partsch
  • Bredde:
    5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.