Chisels, Aesculap®
Chisel, Partsch, 140 mm, 4 mm
Katalog # AESCFL174R
Leverandør: Aesculap
Gå til:
Product Details & Documents
These chisels are used for dental surgery.
Specifikationer
- Beskrivelse:Chisel
- Længde:140 mm
- Seq:06
- Model:Partsch
- Bredde:4 mm