Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Chisels, Aesculap®

Chisels, Aesculap®

Chisel, angled, -, 140 mm, 12 mm
Catalog # AESCFL128R
Supplier:   Aesculap
Chisels, Aesculap®
Catalog # AESCFL128R
Supplier:  Aesculap
Chisels, Aesculap®

Chisel, angled, -, 140 mm, 12 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

These chisels are used for dental surgery.

Specifications
  • Beskrivelse:
    Chisel, angled
  • Længde:
    140 mm
  • Seq:
    23
  • Model:
    -
  • Bredde:
    12 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.