Orders LinkContactUsLinkComponent
Bone curettes, Aesculap®
Bone curettes, Aesculap®
Bone curette, type Halle (Fig. 1), with 6,8 mm end, flexible shaft, 218 mm
Katalog # AESCFK686R
Leverandør:   Aesculap
Bone curettes, Aesculap®
Katalog # AESCFK686R
Leverandør:  Aesculap
Bone curettes, Aesculap®
Bone curette, type Halle (Fig. 1), with 6,8 mm end, flexible shaft, 218 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Bone curette, type Halle (Fig. 1), with 6,8 mm end, flexible shaft
  • Længde:
    218 mm
  • Seq:
    41