Orders LinkContactUsLinkComponent
Bone curettes, Aesculap®
Bone curettes, Aesculap®
Bone curette, type Halle (Fig. 00), with 4,4 mm end, flexible shaft, 215 mm
Catalog # AESCFK684R
Supplier:   Aesculap
Bone curettes, Aesculap®
Catalog # AESCFK684R
Supplier:  Aesculap
Bone curettes, Aesculap®
Bone curette, type Halle (Fig. 00), with 4,4 mm end, flexible shaft, 215 mm
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Bone curette, type Halle (Fig. 00), with 4,4 mm end, flexible shaft
  • Længde:
    215 mm
  • Seq:
    39