Bone curettes, Aesculap®
Bone curette, type Lempert (Fig. 0000), with 1,8 mm end, 215 mm
AESCFK675R
: Aesculap
- Beskrivelse:Bone curette, type Lempert (Fig. 0000), with 1,8 mm end
- Længde:215 mm
- Seq:34
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Natriumhydroxid 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals