Bone curettes, Aesculap®
Bone curette, type Daubenspeck (Fig. 000), with 3,6 mm end, angled upwards, 200 mm
Katalog # AESCFK663R
Leverandør: Aesculap
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Bone curette, type Daubenspeck (Fig. 000), with 3,6 mm end, angled upwards
- Længde:200 mm
- Seq:31