Bone curettes, Aesculap®
Bone curette, Volkmann type (Fig. 1), with 6,8 mm end, 170 mm
AESCFK634R
: Aesculap
- Beskrivelse:Bone curette, Volkmann type (Fig. 1), with 6,8 mm end
- Længde:170 mm
- Seq:19