Bone curettes, Aesculap®
Bone curette, Bruns type (Fig. 0), with 5 mm end, 165 mm
Katalog # AESCFK613R
Leverandør: Aesculap
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Bone curette, Bruns type (Fig. 0), with 5 mm end
- Længde:165 mm
- Seq:03