Dilator set, Aesculap®
Dilator, 7 mm
Catalog # AESCEM107R
Supplier: Aesculap
Jump to:
Product Details & Documents
Complete set of uterine dilators.
Delivery information: Consists of 26 dilators, 1 rack for dilators (1 to 17 mm), 1 rack for dilators (18 to 26 mm) and metal case with hinged aluminium lid (330×220×80 mm).
Specifications
- Beskrivelse:Dilator, 7 mm
- Seq:08