Chisels, Aesculap®
Chisel, straight, with PEEK handle, -, 200 mm, 20 mm
Katalog # AESCDO754R
Leverandør: Aesculap
Gå til:
Product Details & Documents
These chisels are used for dental surgery.
Specifikationer
- Beskrivelse:Chisel, straight, with PEEK handle
- Længde:200 mm
- Seq:29
- Model:-
- Bredde:20 mm
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



