Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Chisels, Aesculap®

Chisels, Aesculap®

Chisel, curved, with PEEK handle, -, 200 mm, 10 mm
Katalog # AESCDO751R
Leverandør:   Aesculap
Chisels, Aesculap®
Katalog # AESCDO751R
Leverandør:  Aesculap
Chisels, Aesculap®

Chisel, curved, with PEEK handle, -, 200 mm, 10 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These chisels are used for dental surgery.

Specifikationer
  • Beskrivelse:
    Chisel, curved, with PEEK handle
  • Længde:
    200 mm
  • Seq:
    24
  • Model:
    -
  • Bredde:
    10 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.