Chisels, Aesculap®
Chisel, straight, with PEEK handle, -, 200 mm, 10 mm
Katalog # AESCDO750R
Leverandør: Aesculap
Gå til:
Product Details & Documents
These chisels are used for dental surgery.
Specifikationer
- Beskrivelse:Chisel, straight, with PEEK handle
- Længde:200 mm
- Seq:25
- Model:-
- Bredde:10 mm