Dissecting scissors, Aesculap® Durotip®
Staright, blunt, 145 mm
Katalog # AESCBC258R
Leverandør: Aesculap
Gå til:
Product Details & Documents
Dissecting scissor for dental applications. Type Baby-Metzenbaum.
- With inserted carbide cutting edges
- Excellent cutting accuracy
- Long working life
- Gold-plated rings
Wave cut models prevent tissue from slipping out during the cutting process.
Specifikationer
- Model:Staright, blunt
- Længde:145 mm