Orders LinkContactUsLinkComponent
Dissecting scissors, Aesculap® Durotip®
Dissecting scissors, Aesculap® Durotip®
Staright, blunt, 145 mm
Katalog # AESCBC258R
Leverandør:   Aesculap
Dissecting scissors, Aesculap® Durotip®
Katalog # AESCBC258R
Leverandør:  Aesculap
Dissecting scissors, Aesculap® Durotip®
Staright, blunt, 145 mm
Price
1.920,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Dissecting scissor for dental applications. Type Baby-Metzenbaum.
  • With inserted carbide cutting edges
  • Excellent cutting accuracy
  • Long working life
  • Gold-plated rings
Wave cut models prevent tissue from slipping out during the cutting process.
Specifikationer
  • Model:
    Staright, blunt
  • Længde:
    145 mm