Orders LinkContactUsLinkComponent
Forceps, micro, jeweller pattern, Aesculap®
Forceps, micro, jeweller pattern, Aesculap®
Straight, very fine (0,2 mm), 110 mm
Katalog # AESCBD330R
Leverandør:   Aesculap
Forceps, micro, jeweller pattern, Aesculap®
Katalog # AESCBD330R
Leverandør:  Aesculap
Forceps, micro, jeweller pattern, Aesculap®
Straight, very fine (0,2 mm), 110 mm
Price
229,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Model:
    Straight, very fine (0,2 mm)
  • Længde:
    110 mm
  • Seq:
    02