Orders LinkContactUsLinkComponent
Dissecting scissors, Aesculap® BipoJet®
Dissecting scissors, Aesculap® BipoJet®
Metzenbaum, standard tip, 230 mm
Katalog # AESCBC687R
Leverandør:   Aesculap
Dissecting scissors, Aesculap® BipoJet®
Katalog # AESCBC687R
Leverandør:  Aesculap
Dissecting scissors, Aesculap® BipoJet®
Metzenbaum, standard tip, 230 mm
Price
8.030,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Ceramic/stainless steel.
  • Combines dissecting scissors, dissecting forceps and monopolar HF electrode in one instrument: No instrument change required
  • Ceramic blade for improved sharpness and longer life, ergonomic handle
  • Special ceramic components are used for the iinstruments to insulate the working area
  • Low thermal spread in the coagulation zone and the consequent avoidance of collateral coagulation damage
These innovative instruments for open surgery combine cutting and coagulation in one step: The tissue is cut while coagulation mode of the HF generator is activated. Prior to performance of the cut, the scissors are rotated so as to coagulate more robust tissue and larger blood vessels. Bleeding can be precisely controlled in localized areas with the scissors tip closed or slightly open. Surface coagulation is performed between the open blades of the scissors.
Specifikationer
  • Model:
    Metzenbaum, standard tip
  • Længde:
    230 mm
  • Seq:
    02