Dissecting forceps, Overholt type, Aesculap®
Fine pattern, figure 1, 210 mm
Katalog # AESCBJ081R
Leverandør: Aesculap
Gå til:
Product Details & Documents
Curved dissecting and ligature forceps. Different figures available.
Specifikationer
- Model:Fine pattern, figure 1
- Længde:210 mm
- Seq:03