Laboratoriesakse
Spids/stump, 165 mm
Katalog # BOCH4005
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Straight scissors made of stainless steel.
Advarsel: These scissors are not recommended for autoclaving.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Model:Spids/stump
- Længde:165 mm
- Seq:06