Wire baskets, square and rectangular
100×100×100 mm
Katalog # BOCH10020
Leverandør: BOCHEM
Gå til:
Product Details & Documents
These stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
- Spot-welded grid 8×8×1 mm
- Reinforced edges
- Very good chemical and temperature resistance
Specifikationer
- B×D×H:100×100×100 mm