Orders LinkContactUsLinkComponent
Bægerglas, lav type
500 ml, 110 mm, 80 mm
Katalog # BOCH8502
Leverandør:   BOCHEM
Bægerglas, lav type
Katalog # BOCH8502
Leverandør:  BOCHEM
Bægerglas, lav type
Bægerglas, lav type
500 ml, 110 mm, 80 mm
Price
839,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel, 18/10.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Specifikationer
  • Udv.Ø:
    80 mm
  • Kapacitet:
    500 ml
  • Højde:
    110 mm