Orders LinkContactUsLinkComponent
Pots, laboratory, without lid
Pots, laboratory, without lid
Lids for laboratory pots, stainless steel 18/10, for 10,5 l pots
Katalog # BOCH8375
Leverandør:   BOCHEM
Pots, laboratory, without lid
Katalog # BOCH8375
Leverandør:  BOCHEM
Pots, laboratory, without lid
Pots, laboratory, without lid
Lids for laboratory pots, stainless steel 18/10, for 10,5 l pots
Price
403,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel 18/10.
  • Rounded corners for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
Multipurpose pots for a wide range of uses. This series is suitable for induction fields.
Bestillingsoplysninger: Lids to be ordered separately.
Specifikationer
  • Beskrivelse:
    Lids for laboratory pots, stainless steel 18/10, for 10,5 l pots
  • Seq:
    14