Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Bowls

100 ml, 40 mm, 80 mm
  BOCH8670
 :   BOCHEM
Bowls
  BOCH8670
 :  BOCHEM
Bowls
Bowls

100 ml, 40 mm, 80 mm

 
226,00 kr
   
1 * 1 Items
 

Made of stainless steel with flat bottom.

  • Very good temperature resistance
  • Very good chemical resistance
  • Easy to clean
 
  • Udv.Ø:
    80 mm
  • Kapacitet:
    100 ml
  • Højde:
    40 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.