Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tænger til digler, robust type

Tænger til digler, robust type

500 mm, 35 mm
Catalog # BOCH2145
Supplier:   BOCHEM
Tænger til digler, robust type
Catalog # BOCH2145
Supplier:  BOCHEM
Tænger til digler, robust type
Tænger til digler, robust type

500 mm, 35 mm

Price
318,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Crucible tongs made from electrolytically polished stainless steel.

Specifications
  • Længde:
    500 mm
  • Seq:
    06
  • Dækket område:
    35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.