Laboratory tables, without ground plate
Laboratoriearbejdsbord, med venstre og højre skuffer, 2000×750×750 mm
Katalog # BOCH11502
Leverandør: BOCHEM
Gå til:
Product Details & Documents
These laboratory tables are made up of 18/10 stainless steel and height adjustable.
- Drawer face with user friendly handle bar
- Noise protection
- Worktops underlaid with soundproofing acoustic wood
Working plate with matt finishing and corners are continuously welded and polished.
Drawer with guard rails made of stainless steel to mount GN-drawer. Drawers can be taken off for cleaning, but are secured against drawing out.
Specifikationer
- Beskrivelse:Laboratoriearbejdsbord, med venstre og højre skuffer
- B×D×H:2000×750×750 mm