Spatulas, knob end
210 mm, 70 mm, 11 mm
Katalog # 231-1055
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Stainless steel. One sided spatulas, available individually or in economical packs of five.
- Heat and chemical resistant
Specifikationer
- Længde:210 mm
- Bladbredde:11 mm
- Klinge længde:70 mm