Spatulas, knob end
150 mm, 60 mm, 9 mm
Katalog # 231-1054
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Stainless steel. One sided spatulas, available individually or in economical packs of five.
- Heat and chemical resistant
Specifikationer
- Længde:150 mm
- Bladbredde:9 mm
- Klinge længde:60 mm