Spatulas, knob end
300 mm, 75 mm, 16 mm
Katalog # 231-1056
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Stainless steel. One sided spatulas, available individually or in economical packs of five.
- Heat and chemical resistant
Specifikationer
- Længde:300 mm
- Bladbredde:16 mm
- Klinge længde:75 mm