Orders LinkContactUsLinkComponent
Laboratory tables, without ground plate
Laboratory tables, without ground plate
Laboratoriearbejdsbord, med venstre og højre skuffer, 2500×750×900 mm
Catalog # BOCH11303
Supplier:   BOCHEM
Laboratory tables, without ground plate
Catalog # BOCH11303
Supplier:  BOCHEM
Laboratory tables, without ground plate
Laboratory tables, without ground plate
Laboratoriearbejdsbord, med venstre og højre skuffer, 2500×750×900 mm
Price
26.630,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These laboratory tables are made up of 18/10 stainless steel and height adjustable.
  • Drawer face with user friendly handle bar
  • Noise protection
  • Worktops underlaid with soundproofing acoustic wood
Working plate with matt finishing and corners are continuously welded and polished.
Drawer with guard rails made of stainless steel to mount GN-drawer. Drawers can be taken off for cleaning, but are secured against drawing out.
Specifications
  • Beskrivelse:
    Laboratoriearbejdsbord, med venstre og højre skuffer
  • B×D×H:
    2500×750×900 mm