Orders LinkContactUsLinkComponent
Laboratory tables, without ground plate
Laboratory tables, without ground plate
Laboratoriearbejdsbord, med venstre og højre skuffer, 2000×750×900 mm
Katalog # BOCH11302
Leverandør:   BOCHEM
Laboratory tables, without ground plate
Katalog # BOCH11302
Leverandør:  BOCHEM
Laboratory tables, without ground plate
Laboratory tables, without ground plate
Laboratoriearbejdsbord, med venstre og højre skuffer, 2000×750×900 mm
Price
24.620,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These laboratory tables are made up of 18/10 stainless steel and height adjustable.
  • Drawer face with user friendly handle bar
  • Noise protection
  • Worktops underlaid with soundproofing acoustic wood
Working plate with matt finishing and corners are continuously welded and polished.
Drawer with guard rails made of stainless steel to mount GN-drawer. Drawers can be taken off for cleaning, but are secured against drawing out.
Specifikationer
  • Beskrivelse:
    Laboratoriearbejdsbord, med venstre og højre skuffer
  • B×D×H:
    2000×750×900 mm