Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Carbon standards

Carbon standards

Organic carbon standard, 50 ppm, 500 mL
Katalog # REGETOC50
Leverandør:   REAGECON
Carbon standards
Katalog # REGETOC50
Leverandør:  REAGECON
CAS nummer:  
Carbon standards

Organic carbon standard, 50 ppm, 500 mL

CAS nummer:  
Price
kr1,540.00
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.

These standards are independent, traceable and certified.

Certifications: Accredited to IEC/ISO 17025 and ISO 17034.

Documents

Specifikationer

Specifikationer

  • Koncentration:
    50 ppm
  • Pakn:
    500 mL
  • Beskrivelse:
    Organic carbon standard

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.