Orders LinkContactUsLinkComponent
Diffraction/transmission gratings
Diffraction/transmission gratings
Transmission grating 600 lines/mm
Katalog # BINH1003079
Marketsource
Leverandør:   3B Scientific
Diffraction/transmission gratings
Katalog # BINH1003079
Leverandør:  3B Scientific
Marketsource
Diffraction/transmission gratings
Diffraction/transmission gratings
Transmission grating 600 lines/mm
Price
925,99 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Transmission grating for spectroscopic examinations and for experiments on diffraction and interference. Suitable to resolve the sodium D lines.
  • Mounted on glass carrier
  • Dimensions: 38×50 mm
Specifikationer
  • Beskrivelse:
    Transmission grating 600 lines/mm