Instrument trays
208×268×17 mm
Katalog # BURK4230-3027
Leverandør: BURKLE
Gå til:
Product Details & Documents
Melamine, white.
Tray without lid
Suitable for use with foodstuffs
Not autoclavable
Not suitable for high temperatures
Bestillingsoplysninger: Lid available as accessory
Specifikationer
- B×D×H:208×268×17 mm