Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for empty columns C series

Accessories for empty columns C series

Net rings for columns, 16 mm I-Ø, 10 µm
Katalog # 18-8761-01
Leverandør:   Cytiva
Accessories for empty columns C series
Katalog # 18-8761-01
Leverandør:  Cytiva
Accessories for empty columns C series
Accessories for empty columns C series

Net rings for columns, 16 mm I-Ø, 10 µm

Price
475,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Net rings for columns, 16 mm I-Ø, 10 µm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.