Orders LinkContactUsLinkComponent
Universal sakse
Universal sakse
Sharp, 130 mm
Katalog # 231-1292
Leverandør:   BOCHEM
Universal sakse
Katalog # 231-1292
Leverandør:  BOCHEM
Universal  sakse
Universal sakse
Sharp, 130 mm
Price
54,30 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel. Multi-purpose scissors for a wide range of applications.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling
Specifikationer
  • Model:
    Sharp
  • Længde:
    130 mm
  • Seq:
    04