Accessories for Corning CellSTACK culture chambers
Vent cap with tubing and 50 mm hydrophobic PVDF 0,2 µm vent filter; Int. Ø: 9,525 mm; Tubing length: 69,85 mm
Leverandør: Corning
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Vent cap with tubing and 50 mm hydrophobic PVDF 0,2 µm vent filter; Int. Ø: 9,525 mm; Tubing length: 69,85 mm