Orders LinkContactUsLinkComponent
Dichlormethan ≥98% stabilised, TECHNICAL
Dichlormethan ≥98% stabilised, TECHNICAL
Plastic bottle, fluorinated, 5 L
Katalog # 25629.364
Sundhed
Leverandør:   VWR Chemicals
Dichlormethan ≥98% stabilised, TECHNICAL
Katalog # 25629.364
Leverandør:  VWR Chemicals
CAS nummer:  75-09-2
Dichlormethan ≥98% stabilised, TECHNICAL
Dichlormethan ≥98% stabilised, TECHNICAL
Plastic bottle, fluorinated, 5 L
CAS nummer:  75-09-2
Price
1.030,00 kr
Log ind for tilgængelighed
1 * 5 L
Product Details & Documents
Synonymer:  Methylenchlorid
  • CAS nummer:75-09-2
  • Formel:CH₂Cl₂
  • MDL nummer:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Kogende Pt:39,8 °C (1013 hPa)
  • Smeltende Pt:–95 °C
  • Tæthed:1,322 g/cm³ (20 °C)
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :stabiliseret med ethanol 0,15 %
  • Opbevaringstemperatur:Stue temperatur
  • FN:1593
  • NÅ:01-2119480404-41
Specifikationer
Specifikationstestresultater
  • Assay (on anhydrous substance):
    Min. 98 %
Specifikationer
  • Pakning:
    Plastic bottle, fluorinated
  • Pakn:
    5 L