Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ethylenglycol ≥98%, TECHNICAL

Ethylenglycol ≥98%, TECHNICAL

Plastic drum, 25 L
Katalog # 24407.463
Irriterende
   Sikrere sundhed for mennesker og miljø
Leverandør:   VWR Chemicals
Ethylenglycol ≥98%, TECHNICAL
Katalog # 24407.463
Leverandør:  VWR Chemicals
   Sikrere sundhed for mennesker og miljø
CAS nummer:  107-21-1
Ethylenglycol ≥98%, TECHNICAL
Ethylenglycol ≥98%, TECHNICAL

Plastic drum, 25 L

CAS nummer:  107-21-1
Price
3.410,00 kr
Log ind for tilgængelighed
1 * 25 L
Product Details & Documents
Synonymer:  1,2-Ethandiol, Glykol
  • CAS nummer:107-21-1
  • Formel:HOCH₂CH₂OH
  • MDL nummer:MFCD00002885
  • Molecular Weight:62.07 g/mol
  • Merck Index:13,03832
  • Kogende Pt:196…198 °C (1013 hPa)
  • Smeltende Pt:–13 °C
  • Tæthed:1,115 g/cm³ (20 °C)
  • Flash Pt:116 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-473-3
  • NÅ:01-2119456816-28
Specifikationer

Specifikationstestresultater

  • Assay (on anhydrous substance):
    Min. 98 %
  • Appearance of solution (50 % V/V; water):
    Passes test

Specifikationer

  • Pakning:
    Plastic drum
  • Pakn:
    25 L
  • Cat. No.:
    24407.463
  • Miljøcertificering:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Sikrere sundhed for mennesker og miljø

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.