Ethylenglycol ≥98%, TECHNICAL
Plastic bottle, 5 L
Leverandør: VWR Chemicals
Ethylenglycol ≥98%, TECHNICAL
Katalog # 24407.361
Leverandør: VWR Chemicals
CAS nummer: 107-21-1
Gå til:
Product Details & Documents
Synonymer: 1,2-Ethandiol, Glykol
- CAS nummer:107-21-1
- Formel:HOCH₂CH₂OH
- MDL nummer:MFCD00002885
- Molecular Weight:62.07 g/mol
- Merck Index:13,03832
- Kogende Pt:196…198 °C (1013 hPa)
- Smeltende Pt:–13 °C
- Tæthed:1,115 g/cm³ (20 °C)
- Flash Pt:116 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-473-3
- NÅ:01-2119456816-28
Certifikater
Søg efter partinummer1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Specifikationer
Specifikationstestresultater
- Assay (on anhydrous substance):Min. 98 %
- Appearance of solution (50 % V/V; water):Passes test
Specifikationer
- Pakning:Plastic bottle
- Pakn:5 L
- Cat. No.:24407.361
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



