Accessories for heating circulators
2× adapters, M16×1 female to NPT ¼" male
Catalog # JULA8890004
Supplier: JULABO GmbH
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:2× adapters, M16×1 female to NPT ¼" male