Spore strips, Attest™
Bacillus stearothermophilus-sporer/strip, dampsterilisation
Leverandør: 3M MEDICAL
Gå til:
Product Details & Documents
3M™ Attest™ Biological Monitoring Systems are true biological indicators, in self-contained indicator capsules. They enable checks on the biological reliability and performance of each steriliser without laboratory facilities and laboratory trained personnel.
- Self-contained, reducing the risk of false positives due to contamination
- Colour change gives clear unambiguous, visual indication of PASS or FAIL
- Time saving and results in 24 to 48 hours which require no interpretation
Documents
Specifikationer
- Beskrivelse:Bacillus stearothermophilus-sporer/strip, dampsterilisation