H-type gel permeation columns, TSK-Gel®
HHR-L, Forkolonne, 6,00 mm, 40 mm
Katalog # 554-1638
Leverandør: TOSOH Bioscience
Gå til:
Product Details & Documents
Stainless steel columns on polymer basis.
Specifikationer
- Kolonne form:Forkolonne
- Ø indv.:6,00 mm
- Beskrivelse:HHR-L
- Længde:40 mm
- Seq:14