Hydrophobic interaction chromatography columns, TSK-GEL®
Guardgel kit, for P/Ns 14013 and 14014, Forkolonne, Ether-5PW, 20 µm
Katalog # TOYO0014025
Leverandør: TOSOH Bioscience
Hydrophobic interaction chromatography columns, TSK-GEL®
Katalog # TOYO0014025
Leverandør: TOSOH Bioscience
Gå til:
Product Details & Documents
These are polymethacrylate-based columns with a choice of three ligands (ether, phenyl or butyl) with varied hydrophobicities from low to high. Suitable for LC-MS applications.
- Rigid polymeric base resin
- Eliminates undesirable interactions with column hardware
- Wide pH range (2 to 12) enables robust cleaning options
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Kolonne form:Forkolonne
- Partikelstørrelse:20 µm
- Beskrivelse:Guardgel kit, for P/Ns 14013 and 14014
- Fase:Ether-5PW