H-type gel permeation columns, TSK-Gel®
HXL-H, Forkolonne, 6,00 mm, 40 mm
554-1439
Stainless steel columns on polymer basis.
- Kolonne form:Forkolonne
- Ø indv.:6,00 mm
- Beskrivelse:HXL-H
- Længde:40 mm
- Seq:13
Stainless steel columns on polymer basis.