Hydrophobic interaction chromatography columns, TSK-GEL®
Phenyl-5PW, Analytisk, Phenyl-5PW, 1000 Å, 10 µm, 5,00 mm, 50 mm
Catalog # TOYO0013063
Supplier: TOSOH Bioscience
Hydrophobic interaction chromatography columns, TSK-GEL®
Catalog # TOYO0013063
Supplier: TOSOH Bioscience
Jump to:
Product Details & Documents
These are polymethacrylate-based columns with a choice of three ligands (ether, phenyl or butyl) with varied hydrophobicities from low to high. Suitable for LC-MS applications.
- Rigid polymeric base resin
- Eliminates undesirable interactions with column hardware
- Wide pH range (2 to 12) enables robust cleaning options
Documents
Certificates
Search by Lot NumberSpecifications
- Kolonne form:Analytisk
- Ø indv.:5,00 mm
- Partikelstørrelse:10 µm
- Porestørrelse:1000 Å
- Beskrivelse:Phenyl-5PW
- Længde:50 mm
- Fase:Phenyl-5PW