Cypermethrin
Cypermethrin
Katalog # EHERC11890000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CC1(C)C(C=C(Cl)Cl)C1C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2
- Inchi: InChI=1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3
- IUPAC: [cyano-(3-phenoxyphenyl)methyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:Cypermethrin
- UN No.:3349
- Cat. No.:EHERC11890000
- CAS-nr:52315-07-8
- Molekylvægt:415,07
- Fareklassebeskrivelse:GHS06, GHS07, GHS08, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Shipping conditions:RT
- Formel:C₂₂H₁₉Cl₂NO₃
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



