Orders LinkContactUsLinkComponent
PLOT columns, CP-PoraPLOT U and CP-PoraPLOT S
PLOT columns, CP-PoraPLOT U and CP-PoraPLOT S
Kapillær, 8,00 µm, 0,25 mm, 25 m
Katalog # VARICP7579
Leverandør:   VARIAN
PLOT columns, CP-PoraPLOT U and CP-PoraPLOT S
Katalog # VARICP7579
Leverandør:  VARIAN
PLOT columns, CP-PoraPLOT U and CP-PoraPLOT S
PLOT columns, CP-PoraPLOT U and CP-PoraPLOT S
Kapillær, 8,00 µm, 0,25 mm, 25 m
Price
7.820,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
CP-PoraPLOT U and CP-PoraPLOT S provide symmetrical peaks from polar and non-polar volatiles for ultimate accuracy.
  • Minimal particle loss reduces detector spiking for reliable results
  • Repeatable retention times for better long-term stability
CP-PoraPLOT U provides symmetrical peaks with polar volatiles. Water has no effect on retention times and elutes as a sharp and quantifiable peak. CP-PoraPLOT U is the most polar porous polymer PLOT column and is designed for halogenated compounds, hydrocarbons C1-C6, ketones and solvents.
CP-PoraPLOT S is a divinylbenzene/vinylpyridine polymer for hydrocarbons and ketones. This phase is ideal for the analysis of medium polarity volatiles, including hydrocarbons and ketones, at higher temperatures than CP-PoraPLOT U.
Specifikationer
  • Kolonne form:
    Kapillær
  • Filmtykkelse:
    8,00 µm
  • Ø indv.:
    0,25 mm
  • Længde:
    25 m
  • Seq:
    01