Orders LinkContactUsLinkComponent
Redox standards
Redox standards
Standard redox solution, 600 mV at 25 °C, 500 mL
Katalog # REGERS600
Leverandør:   REAGECON
Redox standards
Katalog # REGERS600
Leverandør:  REAGECON
CAS nummer:  
Redox standards
Standard redox solution, 600 mV at 25 °C, 500 mL
CAS nummer:  
Price
939,00 kr
Log ind for tilgængelighed
1 * 500 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
The potentials quoted are of platinum electrode with reference to Ag/AgCl (3 M KCl).
Documents
Specifikationer
Specifikationer
  • Pakn:
    500 mL
  • Beskrivelse:
    Standard redox solution, 600 mV at 25 °C
Specifikationer